4-(N,N-Diphenylamino)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Diphenylaminobenzaldehyde

AS33133G1 AS33133G10 AS33133G5
ACTIAS33133G1EA 0 CHF
ACTIAS33133G1 ACTIAS33133G10 ACTIAS33133G5
4-(N,N-Diphenylamino)benzaldehyd
4-(N,N-Diphenylamino)benzaldehyd
Formel: C₁₉H₁₅NO
MDL Number: MFCD00145131
CAS-Nummer: 4181-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR