4-Brom-2-(methylsulfonyl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS22456G1 AS22456G25 AS22456G5
ACTIAS22456G1EA 129 CHF
ACTIAS22456G1 ACTIAS22456G25 ACTIAS22456G5
4-Brom-2-(methylsulfonyl)benzaldehyd
4-Bromo-2-(methylsulphonyl)benzaldehyde
Formel: C₈H₇BrO₃S
MW: 263,11 g/mol
MDL Number: MFCD04037947
CAS-Nummer: 849035-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR