3-Brom-4-(methylsulfonyl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS22463G1 AS22463G5
ACTIAS22463G1EA 0 CHF
ACTIAS22463G1 ACTIAS22463G5
3-Brom-4-(methylsulfonyl)benzaldehyd
3-Bromo-4-(methylsulphonyl)benzaldehyde
Formel: C₈H₇BrO₃S
MDL Number: MFCD13191600
CAS-Nummer: 254878-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR