4-(1,3-Oxazol-5-yl)anilin

Lieferant: ACTIVATE SCIENTIFIC

AS20800G1 AS20800G25 AS20800G5
ACTIAS20800G1EA 0 CHF
ACTIAS20800G1 ACTIAS20800G25 ACTIAS20800G5
4-(1,3-Oxazol-5-yl)anilin
4-(1,3-Oxazol-5-yl)anilin
Formel: C₉H₈N₂O
MDL Number: MFCD00052192
CAS-Nummer: 1008-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR