2-Chlor-5-pyrimidinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloropyrimidine-5-boronic acid

AS31551G1 AS31551G5
ACTIAS31551G1EA 0 CHF
ACTIAS31551G1 ACTIAS31551G5
2-Chlor-5-pyrimidinboronsäure
2-Chloro-5-pyrimidineboronic acid
Formel: C₄H₄BClN₂O₂
MW: 158.35 g/mol
MDL Number: MFCD08063113
CAS-Nummer: 1003845-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR