2,4-Dimethoxy-5-pyrimidinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51290G1 AS51290G5
ACTIAS51290G1EA 0 CHF
ACTIAS51290G1 ACTIAS51290G5
2,4-Dimethoxy-5-pyrimidinboronsäure
2,4-Dimethoxy-5-pyrimidineboronic acid
Formel: C₆H₉BN₂O₄
MDL Number: MFCD01318985
CAS-Nummer: 89641-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR