2-Ethoxy-5-pyrimidinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-Ethoxypyrimidin-5-yl)boronic acid

AS51245G1 AS51245G5
ACTIAS51245G1EA 0 CHF
ACTIAS51245G1 ACTIAS51245G5
2-Ethoxy-5-pyrimidinboronsäure
2-Ethoxy-5-pyrimidineboronic acid
Formel: C₆H₉BN₂O₃
MW: 167.9600 g/mol
MDL Number: MFCD07375134
CAS-Nummer: 1003043-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR