3-Chlor-4-(1H-pyrazol-1-yl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Chloro-4-(1-pyrazolyl)benzaldehyde

AS21508G1 AS21508G5
ACTIAS21508G1EA 164 CHF
ACTIAS21508G1 ACTIAS21508G5
3-Chlor-4-(1H-pyrazol-1-yl)benzaldehyd
3-Chlor-4-(1H-pyrazol-1-yl)benzaldehyd
Formel: C₁₀H₇ClN₂O
MW: 206,6284 g/mol
MDL Number: MFCD11656795
CAS-Nummer: 1186663-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR