3-Brom-4-(1H-pyrazol-1-yl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS21543G1 AS21543G5
ACTIAS21543G1EA 220 CHF
ACTIAS21543G1 ACTIAS21543G5
3-Brom-4-(1H-pyrazol-1-yl)benzaldehyd
3-Brom-4-(1H-pyrazol-1-yl)benzaldehyd
Formel: C₁₀H₇BrN₂O
MDL Number: MFCD11656798
CAS-Nummer: 1186663-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR