5-Iod-1-methyl-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Iodo-1-methylpyrazole

AS32349G1 AS32349G5
ACTIAS32349G1EA 0 CHF
ACTIAS32349G1 ACTIAS32349G5
5-Iod-1-methyl-1H-pyrazol
5-Iodo-1-methyl-1H-pyrazole
Formel: C₄H₅IN₂
MW: 208,00 g/mol
MDL Number: MFCD11109327
CAS-Nummer: 34091-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR