4-Iod-3-methyl-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

AS34838G1 AS34838G5
ACTIAS34838G1EA 0 CHF
ACTIAS34838G1 ACTIAS34838G5
4-Iod-3-methyl-1H-pyrazol
4-Iodo-3-methyl-1H-pyrazole
Formel: C₄H₅IN₂
MDL Number: MFCD09908176
CAS-Nummer: 15802-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR