3-Iod-1-methyl-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

AS15076G1 AS15076G5
ACTIAS15076G1EA 0 CHF
ACTIAS15076G1 ACTIAS15076G5
3-Iod-1-methyl-1H-pyrazol
3-Iodo-1-methyl-1H-pyrazole
Formel: C₄H₅IN₂
MW: 208.00 g/mol
MDL Number: MFCD08435890
CAS-Nummer: 92525-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR