1-Ethyl-3-iod-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

AS15309G1 AS15309G5
ACTIAS15309G1EA 0 CHF
ACTIAS15309G1 ACTIAS15309G5
1-Ethyl-3-iod-1H-pyrazol
1-Ethyl-3-iodo-1H-pyrazole
Formel: C₅H₇IN₂
MDL Number: MFCD12756564
CAS-Nummer: 1202781-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR