4-Brom-3-methyl-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-3-methylpyrazole

AS23178G100 AS23178G25
ACTIAS23178G100EA 141 CHF
ACTIAS23178G100 ACTIAS23178G25
4-Brom-3-methyl-1H-pyrazol
4-Bromo-3-methyl-1H-pyrazole
Formel: C₄H₅BrN₂
MW: 161,01 g/mol
MDL Number: MFCD00005241
CAS-Nummer: 13808-64-5
EINECS: 237-456-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR