6-O-Palmitoyl-L-ascorbinsäure
Lieferant: EHRENSTORFER GMBH
Synonyms:
L(+)-Ascorbylpalmitat
, 6-palmitoyl-L-ascorbic acid
EHERC10303930EA
79.2
CHF
EHERC10303930
6-O-Palmitoyl-L-ascorbinsäure
6-O-Palmitoyl-L-ascorbinsäure
Formel:
C₂₂H₃₈O₇ MW: 414,54 g/mol Siedepunkt: ∼250 °C Melting Pt: 107…117 °C |
MDL Number:
MFCD00005377 CAS-Nummer: 137-66-6 EINECS: 205-305-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...