1,4-Benzodioxan-6-boronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2,3-Dihydro-1,4-benzodioxine-6-boronic acid
AS50224G1
AS50224G5
ACTIAS50224G1EA
0
CHF
ACTIAS50224G1
ACTIAS50224G5
1,4-Benzodioxan-6-boronsäure
1,4-Benzodioxan-6-boronsäure
Formel:
C₈H₉BO₄ MW: 179,97 g/mol |
MDL Number:
MFCD01009696 CAS-Nummer: 164014-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...