(R)-1-Boc-3-hydroxypiperidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (R)-1-BOC-3-hydroxypiperidine, (R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate

AS35179G1 AS35179G5
ACTIAS35179G1EA 0 CHF
ACTIAS35179G1 ACTIAS35179G5
(R)-1-Boc-3-hydroxypiperidin
(R)-1-Boc-3-hydroxypiperidin
Formel: C₁₀H₁₉NO₃
MW: 201,27 g/mol
MDL Number: MFCD04115306
CAS-Nummer: 143900-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR