1-Boc-4-(methylamino)piperidin

Lieferant: ACTIVATE SCIENTIFIC

AS31170G1 AS31170G5
ACTIAS31170G1EA 0 CHF
ACTIAS31170G1 ACTIAS31170G5
1-Boc-4-(methylamino)piperidin
1-Boc-4-(methylamino)piperidin
Formel: C₁₁H₂₂N₂O₂
MDL Number: MFCD02259411
CAS-Nummer: 147539-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR