1-Boc-3-(methylamino)piperidin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-(Methylamino)piperidine, N1-BOC protected, 1-Boc-3-methylaminopieridine
AS3108G1
AS3108G5
ACTIAS3108G1EA
0
CHF
ACTIAS3108G1
ACTIAS3108G5
1-Boc-3-(methylamino)piperidin
1-Boc-3-(methylamino)piperidin
Formel:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol |
MDL Number:
MFCD04114980 CAS-Nummer: 392331-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...