1-Boc-3-(ethylamino)piperidin

Lieferant: ACTIVATE SCIENTIFIC

AS3010G1 AS3010G5
ACTIAS3010G1EA 0 CHF
ACTIAS3010G1 ACTIAS3010G5
1-Boc-3-(ethylamino)piperidin
1-Boc-3-(ethylamino)piperidin
Formel: C₁₂H₂₄N₂O₂
MDL Number: MFCD08273954
CAS-Nummer: 883546-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR