(±)-3-Amino-1-Boc-piperidin

Lieferant: ACTIVATE SCIENTIFIC

AS33371G1 AS33371G5
ACTIAS33371G1EA 0 CHF
ACTIAS33371G1 ACTIAS33371G5
(±)-3-Amino-1-Boc-piperidin
3-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
MDL Number: MFCD01861219
CAS-Nummer: 184637-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR