4-Amino-3-iod-1H-pyrazolo[3,4-d]pyrimidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine

AS22737G1 AS22737G25 AS22737G5
ACTIAS22737G1EA 0 CHF
ACTIAS22737G1 ACTIAS22737G25 ACTIAS22737G5
4-Amino-3-iod-1H-pyrazolo[3,4-d]pyrimidin
4-Amino-3-iod-1H-pyrazolo[3,4-d]pyrimidin
Formel: C₅H₄N₅I
MW: 261.03 g/mol
MDL Number: MFCD03787931
CAS-Nummer: 151266-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR