Ionomycin

Lieferant: Molekula

MOLE22000466-1MGEA 0 CHF
MOLE22000466-1MG
Ionomycin
Ionomycin
Formel: C₄₁H₇₂O₉
MDL Number: MFCD06798385
CAS-Nummer: 56092-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR