Brom-tris-pyrrolidinophosphoniumhexafluorophosphat

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS34795G100EA 0 CHF
ACTIAS34795G10 ACTIAS34795G100
Brom-tris-pyrrolidinophosphoniumhexafluorophosphat
Brom-tris-pyrrolidinophosphoniumhexafluorophosphat
Formel: C₁₂H₂₄BrF₆N₃P₂
MW: 466,2 g/mol
MDL Number: MFCD00077412
CAS-Nummer: 132705-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR