Benzophenon-[Carbonyl-13C] (98% ¹³C )
Lieferant: CAMBRIDGE ISOTOPE LABS., INC.
CAMBCLM-3253-0.5EA
0
CHF
CAMBCLM-3253-0.5
Benzophenon-[Carbonyl-13C] (98% ¹³C )
Benzophenon-[Carbonyl-13C]
Formel:
C₁₃H₁₀O |
CAS-Nummer:
32488-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...