4-(1-Pyrrolidinyl)pyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Pyrrolidinopyridine
AS21750G100
AS21750G25
ACTIAS21750G100EA
0
CHF
ACTIAS21750G100
ACTIAS21750G25
4-(1-Pyrrolidinyl)pyridin
4-(1-Pyrrolidinyl)pyridin
Formel:
C₉H₁₂N₂ MW: 148.21 g/mol |
MDL Number:
MFCD00006415 CAS-Nummer: 2456-81-7 EINECS: 219-534-2 UN: 2923 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...