4'-Amino-2'-hydroxyacetophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-(4-Amino-2-hydroxyphenyl)ethanone, 1-(4-Amino-2-hydroxyphenyl)ethan-1-one

AS40044G1 AS40044G5
ACTIAS40044G1EA 0 CHF
ACTIAS40044G1 ACTIAS40044G5
4'-Amino-2'-hydroxyacetophenon
4'-Amino-2'-hydroxyacetophenon
Formel: C₈H₉NO₂
MW: 151,1600 g/mol
MDL Number: MFCD00100636
CAS-Nummer: 2476-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR