5-Formyl-2-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50864G1 AS50864G5
ACTIAS50864G1EA 0 CHF
ACTIAS50864G1 ACTIAS50864G5
5-Formyl-2-methoxyphenylboronsäure
5-Formyl-2-methoxyphenylboronsäure
Formel: C₈H₉BO₄
MDL Number: MFCD01319044
CAS-Nummer: 127972-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR