2-(1H-Pyrazol-1-yl)benzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS30059G1
AS30059G5
ACTIAS30059G1EA
0
CHF
ACTIAS30059G1
ACTIAS30059G5
2-(1H-Pyrazol-1-yl)benzaldehyd
2-(1H-Pyrazol-1-yl)benzaldehyd
Formel:
C₁₀H₈N₂O |
MDL Number:
MFCD03086186 CAS-Nummer: 138479-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...