4'-Brom-2'-fluoracetophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-(4-Bromo-2-fluorophenyl)ethanone

AS32821G1 AS32821G5
ACTIAS32821G1EA 0 CHF
ACTIAS32821G1 ACTIAS32821G5
4'-Brom-2'-fluoracetophenon
4'-Brom-2'-fluoracetophenon
Formel: C₈H₆BrFO
MW: 217.04 g/mol
MDL Number: MFCD03411551
CAS-Nummer: 625446-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR