1H-Pyrazol-4-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Pyrazolecarboxylic acid

AS31266G1 AS31266G5
ACTIAS31266G1EA 0 CHF
ACTIAS31266G1 ACTIAS31266G5
1H-Pyrazol-4-carbonsäure
1H-Pyrazole-4-carboxylic acid
Formel: C₄H₄N₂O₂
MW: 112.09 g/mol
MDL Number: MFCD00011558
CAS-Nummer: 37718-11-9
EINECS: 253-641-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR