Fmoc-phenylalaninol

Lieferant: BACHEM BIOCHEMICA

BACE4025445.0001EA 0 CHF
BACE4025445.0001 BACE4025445.0005
Fmoc-phenylalaninol
Fmoc-phenylalaninol
Formel: C₂₄H₂₃NO₃
MW: 373,452 g/mol
MDL Number: MFCD00235941
CAS-Nummer: 129397-83-7
EINECS: 252-661-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR