2-Hydroxy-6-methylnicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Hydroxy-6-methylpyridine-3-carboxylic acid

AS24877G100 AS24877G25
ACTIAS24877G100EA 354 CHF
ACTIAS24877G100 ACTIAS24877G25
2-Hydroxy-6-methylnicotinsäure
2-Hydroxy-6-methylnicotinsäure
Formel: C₇H₇NO₃
MW: 153,14 g/mol
MDL Number: MFCD00006272
CAS-Nummer: 38116-61-9
EINECS: 253-784-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR