2,3-Dimethylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS21469G100 AS21469G25
ACTIAS21469G100EA 0 CHF
ACTIAS21469G100 ACTIAS21469G25
2,3-Dimethylbenzaldehyd
2,3-Dimethylbenzaldehyde
Formel: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00798004
CAS-Nummer: 5779-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR