3-Nitro-5-(trifluormethyl)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32309G1 AS32309G5
ACTIAS32309G1EA 0 CHF
ACTIAS32309G1 ACTIAS32309G5
3-Nitro-5-(trifluormethyl)benzonitril
3-Nitro-5-(trifluormethyl)benzonitril
Formel: C₈H₃F₃N₂O₂
MDL Number: MFCD03412207
CAS-Nummer: 20566-80-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR