5-Amino-2,4-dichlorpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,4-Dichloropyrimidin-5-amine

AS31838G1 AS31838G5
ACTIAS31838G1EA 0 CHF
ACTIAS31838G1 ACTIAS31838G5
5-Amino-2,4-dichlorpyrimidin
5-Amino-2,4-dichloropyrimidine
Formel: C₄H₃Cl₂N₃
MW: 164,00 g/mol
MDL Number: MFCD05662684
CAS-Nummer: 5177-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR