3-Nitro-5-(trifluormethyl)anisol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Methoxy-5-nitrobenzotrifluoride
AS20291G25
AS20291G5
ACTIAS20291G25EA
270
CHF
ACTIAS20291G25
ACTIAS20291G5
3-Nitro-5-(trifluormethyl)anisol
3-Nitro-5-(trifluormethyl)anisol
Formel:
C₈H₆F₃NO₃ MW: 221.13 g/mol |
MDL Number:
MFCD00010446 CAS-Nummer: 328-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...