4-Hydroxynicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS31329G1 AS31329G5
ACTIAS31329G1EA 0 CHF
ACTIAS31329G1 ACTIAS31329G5
4-Hydroxynicotinsäure
4-Hydroxynicotinsäure
Formel: C₆H₅NO₃
MW: 139,11 g/mol
MDL Number: MFCD00040286
CAS-Nummer: 609-70-1
EINECS: 210-197-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR