2-Aminoisonicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-4-pyridinecarboxylic acid, 2-Aminopyridine-4-carboxylic acid

AS1360G100 AS1360G5
ACTIAS1360G100EA 0 CHF
ACTIAS1360G100 ACTIAS1360G5
2-Aminoisonicotinsäure
2-Aminoisonicotinic acid
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
MDL Number: MFCD00233714
CAS-Nummer: 13362-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR