3-Chlor-1,2-benzisoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS1034G1 AS1034G5
ACTIAS1034G1EA 0 CHF
ACTIAS1034G1 ACTIAS1034G5
3-Chlor-1,2-benzisoxazol
3-Chloro-1,2-benzisoxazole
Formel: C₇H₄ClNO
MW: 153,5658 g/mol
MDL Number: MFCD03407314
CAS-Nummer: 16263-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR