2,6-Diacetylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35441G1 AS35441G5
ACTIAS35441G1EA 0 CHF
ACTIAS35441G1 ACTIAS35441G5
2,6-Diacetylpyridin
2,6-Diacetylpyridin
Formel: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00006304
CAS-Nummer: 1129-30-2
EINECS: 214-442-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR