2-Fluor-3-methyl-5-pyridinboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Fluoro-5-methyl-3-pyridylboronic acid, 2-Fluoro-3-methylpyridine-5-boronic acid, (6-Fluoro-5-methylpyridin-3-yl)boronic acid
AS50720G1
AS50720G5
ACTIAS50720G1EA
0
CHF
ACTIAS50720G1
ACTIAS50720G5
2-Fluor-3-methyl-5-pyridinboronsäure
2-Fluor-3-methyl-5-pyridinboronsäure
Formel:
C₆H₇BFNO₂ MW: 154.94 g/mol |
MDL Number:
MFCD03411562 CAS-Nummer: 904326-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...