2-Fluor-4-methyl-5-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-4-methylpyridine-5-boronic acid

AS50721G1 AS50721G5
ACTIAS50721G1EA 0 CHF
ACTIAS50721G1 ACTIAS50721G5
2-Fluor-4-methyl-5-pyridinboronsäure
2-Fluor-4-methyl-5-pyridinboronsäure
Formel: C₆H₇BFNO₂
MW: 154,9400 g/mol
MDL Number: MFCD08741406
CAS-Nummer: 1072944-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR