2-Chlor-3-methyl-5-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-3-methylpyridine-5-boronic acid, 6-Chloro-5-methylpyridine-3-boronic acid, (6-Chloro-5-methylpyridin-3-yl)boronic acid

AS50921G1 AS50921G5
ACTIAS50921G1EA 0 CHF
ACTIAS50921G1 ACTIAS50921G5
2-Chlor-3-methyl-5-pyridinboronsäure
2-Chlor-3-methyl-5-pyridinboronsäure
Formel: C₆H₇BClNO₂
MDL Number: MFCD03411564
CAS-Nummer: 1003043-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR