2-Chlor-4-methyl-5-pyridinboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(6-Chloro-4-methylpyridin-3-yl)boronic acid, 2-Chloro-4-methylpyridine-5-boronic acid
AS33465G1
AS33465G10
AS33465G5
ACTIAS33465G1EA
0
CHF
ACTIAS33465G1
ACTIAS33465G10
ACTIAS33465G5
2-Chlor-4-methyl-5-pyridinboronsäure
2-Chlor-4-methyl-5-pyridinboronsäure
Formel:
C₆H₇BClNO₂ MW: 171,3900 g/mol |
MDL Number:
MFCD07368882 CAS-Nummer: 913836-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...