2-Methyl-3-pyridinboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methylpyridine-3-boronic acid pinacol ester, 2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

AS24052G1 AS24052G5
ACTIAS24052G1EA 50 CHF
ACTIAS24052G1 ACTIAS24052G5
2-Methyl-3-pyridinboronsäurepinakolester
2-Methyl-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
MW: 219,0878 g/mol
MDL Number: MFCD08669594
CAS-Nummer: 1012084-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR