2,6-Dimethoxy-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid

AS50198G1 AS50198G5
ACTIAS50198G1EA 0 CHF
ACTIAS50198G1 ACTIAS50198G5
2,6-Dimethoxy-3-pyridinboronsäure
2,6-Dimethoxy-3-pyridineboronic acid
Formel: C₇H₁₀BNO₄
MW: 182.97 g/mol
MDL Number: MFCD03788239
CAS-Nummer: 221006-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR