2-Ethoxy-5-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Ethoxy-5-pyridylboronic acid, 6-Ethoxy-3-pyridylboronic acid, 2-Ethoxypyridine-5-boronic acid, (6-Ethoxypyridin-3-yl)boronic acid, 6-Ethoxypyridine-3-boronic acid

AS33858G1 AS33858G10 AS33858G5
ACTIAS33858G1EA 0 CHF
ACTIAS33858G1 ACTIAS33858G10 ACTIAS33858G5
2-Ethoxy-5-pyridinboronsäure
2-Ethoxy-5-pyridinboronsäure
Formel: C₇H₁₀BNO₃
MW: 166.97 g/mol
MDL Number: MFCD06201032
CAS-Nummer: 612845-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR