2-Methoxy-4-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methoxy-4-pyridylboronic acid, 2-Methoxypyridine-4-boronic acid, (2-Methoxypyridin-4-yl)boronic acid

AS31354G1 AS31354G5
ACTIAS31354G1EA 0 CHF
ACTIAS31354G1 ACTIAS31354G5
2-Methoxy-4-pyridinboronsäure
2-Methoxy-4-pyridineboronic acid
Formel: C₆H₈BNO₃
MW: 152.9436 g/mol
MDL Number: MFCD07368877
CAS-Nummer: 762262-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR