3,5-Dimethyl-4-isoxazolboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (dimethyl-1,2-oxazol-4-yl)boronic acid

AS50359G1 AS50359G5
ACTIAS50359G1EA 0 CHF
ACTIAS50359G1 ACTIAS50359G5
3,5-Dimethyl-4-isoxazolboronsäure
3,5-Dimethylisoxazole-4-boronic acid
Formel: C₅H₈BNO₃
MW: 140,93 g/mol
MDL Number: MFCD02677945
CAS-Nummer: 16114-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR